EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Synthesis of aryl(di)azinyl ketones through copper- and iron-catalyzed oxidation of the methylene group of aryl(di)azinylmethanes.
PMID 22290611
Abstract
Sustainable Oxidations: an oxidation method to transform aryl(di)azinylmethanes into aryl(di)azinyl ketones is described. Base metals (copper and iron) as catalysts in combination with O(2) as the oxidant are used, which makes this method sustainable. The utility of this method is illustrated by the synthesis of 6-(4-methylbenzoyl)pyridine-2-carbaldehyde, which is an intermediate in the preparation of the drug Acrivastine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0119
Acrivastine, ≥98% (HPLC)
C22H24N2O2
T6764
Triprolidine hydrochloride, ≥99%
C19H22N2 · HCl