EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Meptazinol and ethanol: a fatal intoxication.
PMID 22290756
Abstract
Meptazinol (Meptid(®)) is an analgesic drug that is used to treat mild to moderate pain including postoperative pain, obstetrical pain, and the pain of renal colic. This case reports a death due to the combined effects of meptazinol and alcohol in a man with significant heart disease and alcoholic liver disease. A 57-year-old male was found unresponsive in his bed at home with empty blister packets of meptazinol around him. A general drug screen detected the presence of meptazinol, and caffeine and metabolites, in cardiac blood. Analysis, both quantitative (HPLC-DAD) and qualitative (HPLC-DAD, LC-MS), of meptazinol was carried out. Meptazinol was found at the following concentrations: 15.5 mg/L in unpreserved femoral blood; 18.6 mg/L in preserved (fluoride-oxalate) femoral blood; 52.1 mg/L in unpreserved cardiac blood; 16.8 mg/L in preserved vitreous; 61.7 mg/L in unpreserved urine; and 9.8 g/L in stomach contents. Ethanol, analyzed by headspace GC-FID, was present in preserved (fluoride-oxalate) femoral venous blood, urine, and vitreous at concentrations of 232 mg/100 mL, 297 mg/100 mL, and 192 mg/100 mL, respectively. Death was attributed to meptazinol and ethanol toxicity, with atherosclerotic coronary artery disease as a contributing factor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2824
Meptazinol hydrochloride
C15H23NO· HCl