EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Pharmacokinetic interactions of flunixin meglumine and doxycycline in broiler chickens.
PMID 22292913
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0530
Clonixin, ≥98% (HPLC)
C13H11ClN2O2
Y0000443
Flunixin meglumine, European Pharmacopoeia (EP) Reference Standard
C14H11F3N2O2·C7H17NO5