EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of novel IP agonists via N-aminoethyl cyclic amines prepared by decarboxylative ring-opening reactions.
PMID 22293844
Abstract
An efficient synthesis of a highly potent and selective IP (PGI(2) receptor) agonist that is not structurally analogous to PGI(2) is described. This synthesis is accomplished through the following key steps: Nucleophilic ring-opening of 3-(4-chlorophenyl)-oxazolidin-2-one prepared by a one-pot procedure with 4-piperidinol and selective O-alkylation of 1-(2-(4-chlorophenylamino)ethyl)piperidin-4-ol. The obtained compound is a potent and selective IP agonist displaying a long duration of action.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
128775
4-Hydroxypiperidine, 98%
C5H11NO
378895
Piperonyloyl chloride, 99%
C8H5ClO3