EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Comparison of the spores of Paenibacillus polymyxa prepared at different temperatures.
PMID 22294453
Abstract
Paenibacillus polymyxa SQR-21, which is antagonistic against Fusarium oxysporum, is used as a biocontrol agent and, when mixed with organic substances for solid fermentation, produces a bioorganic fertilizer. The spores of P. polymyxa prepared at different temperatures were characterized with respect to the dipicolinic acid content, heat resistance, fatty acid composition and germination. Spores prepared at 37°C showed higher heat resistance than those prepared at 25 and 30°C. However, the germination rate was negatively correlated with the sporulation temperature. The maximum germination rate of the spores prepared at 25°C was 1.3-times higher than the spores prepared at 30°C. The sporulation temperature thus affects the resistance and germination properties of P. polymyxa spores. These results are useful for the production of improved bio-organic fertilizer.