EMAIL THIS PAGE TO A FRIEND
Cell biology international
Lipid body biogenesis and the role of microtubules in lipid synthesis in Ornithogalum umbellatum lipotubuloids.
PMID 22295975
Abstract
Lipid bodies present in lipotubuloids of Ornithogalum umbellatum ovary epidermis take the form of a lens between leaflets of ER (endoplasmic reticulum) membrane filled with a highly osmiophilic substance. The two enzymes, DGAT1 [DAG (diacylglycerol) acyltransferase 1] and DGAT2 (DAG acyltransferase 2), involved in this process are synthesized on rough ER and localized in the ER near a monolayer surrounding entities like lipid bodies. After reaching the appropriate size, newly formed lipid bodies transform into mature spherical lipid bodies filled with less osmiophilic content. They appear to be surrounded by a half-unit membrane, with numerous microtubules running adjacently in different directions. The ER, no longer continuous with lipid bodies, makes contact with them through microtubules. At this stage, lipid synthesis takes place at the periphery of lipid bodies. This presumption, and a hypothesis that microtubules are involved in lipid synthesis delivering necessary components to lipid bodies, is based on strong arguments: (i) silver grains first appear over microtubules after a short [3H]palmitic acid incubation and before they are observed over lipid bodies; (ii) blockade of [3H]palmitic acid incorporation into lipotubuloids by propyzamide, an inhibitor of microtubule function; and (iii) the presence of gold grains above the microtubules after DGAT1 and DGAT2 reactions, as also near microtubules after an immunogold method that identifies phospholipase D1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442764
Pronamide (Kerb), analytical standard
C12H11Cl2NO
45645
Propyzamide, PESTANAL®, analytical standard
C12H11Cl2NO