EMAIL THIS PAGE TO A FRIEND
Nanoscale
Probing Y-shaped DNA structure with time-resolved FRET.
PMID 22297682
Abstract
Self-assembly based on nucleic acid systems has become highly attractive for bottom-up fabrication of programmable matter due to the highly selective molecular recognition property of biomolecules. In this context, Y-shaped DNA (Y-DNA) provides an effective building block for forming unique self-assembled large-scale architectures. The dimension and growth of the nano- and microstructures depend significantly on the configurational stability of Y-DNA as a building block. Here we present structural studies of Y-DNA systems using a time-resolved FRET (Förster resonance energy transfer) technique. A fluorophore (Alexa 488) and an acceptor (DABCYL) were placed at two different ends of Y-DNA, and the lifetime of the fluorophore was measured to probe the relative distance between the donor and acceptor. Our results confirmed different distances between the arms of the Y-DNA and highlighted the overall structural integrity of the Y-DNA system as a leading building block for molecular self-assembly. Temperature dependent lifetime measurements indicated configurational changes in the overall Y-DNA nanoarchitecture above 40 °C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73225
Butter Yellow, analytical standard
C14H15N3