EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Crystallization and preliminary X-ray diffraction analysis of an L-amino-acid oxidase from Bothrops jararacussu venom.
PMID 22298002
Abstract
Snake-venom L-amino-acid oxidases (SV-LAAOs) trigger a wide range of local and systematic effects, including inhibition of platelet aggregation, cytotoxicity, haemolysis, apoptosis and haemorrhage. These effects mainly arise from the uncontrolled release of the hydrogen peroxide that is produced by the redox reaction involving L-amino acids catalyzed by these flavoenzymes. Taking their clinical relevance into account, few SV-LAAOs have been structurally characterized and the structural determinants responsible for their broad direct and indirect pharmacological activities remain unclear. In this work, an LAAO from Bothrops jararacussu venom (BJu-LAAO) was purified and crystallized. The BJu-LAAO crystals belonged to space group P2(1), with unit-cell parameters a = 66.38, b = 72.19, c = 101.53 Å, β = 90.9°. The asymmetric unit contained two molecules and the structure was determined and partially refined at 3.0 Å resolution.