The Journal of nutrition
Vision changes after spaceflight are related to alterations in folate- and vitamin B-12-dependent one-carbon metabolism.
PMID 22298570
Approximately 20% (7 of 38) of astronauts on International Space Station (ISS) missions have developed measurable ophthalmic changes after flight. This study was conducted to determine if the folate- and vitamin B-12-dependent 1-carbon metabolic pathway is altered in these individuals. Since 2006, we have conducted experiments on the ISS to evaluate nutritional status and related biochemical indices of astronauts before, during, and after flight. Data were modeled to evaluate differences between individuals with ophthalmic changes (n = 5) and those without them (n = 15), all of whom were on ISS missions of 48-215 d. We also determined whether mean preflight serum concentrations of the 1-carbon metabolites and changes in measured cycloplegic refraction after flight were associated. Serum homocysteine (Hcy), cystathionine, 2-methylcitric acid (2MCA), and methylmalonic acid concentrations were 25-45% higher (P < 0.001) in astronauts with ophthalmic changes than in those without them. These differences existed before, during, and after flight. Preflight serum concentrations of Hcy and cystathionine, and mean in-flight serum folate, were correlated with change (postflight relative to preflight) values in refraction (P < 0.05), and preflight serum concentrations of 2MCA tended to be associated (P = 0.06) with ophthalmic changes. The biochemical differences observed in crewmembers with vision issues strongly suggest that their folate- and vitamin B-12-dependent 1-carbon transfer metabolism was affected before and during flight. The consistent differences in markers of 1-carbon metabolism between those who did and those who did not develop changes in vision suggest that polymorphisms in enzymes of this pathway may interact with microgravity to cause these pathophysiologic changes.
Related Materials
Product #
Molecular Formula
Add to Cart
L-Cystathionine, ≥98% (TLC)