EMAIL THIS PAGE TO A FRIEND
Surgical innovation
Ethyl pyruvate improves healing of colonic anastomosis in a rat model of peritonitis.
PMID 22298750
Abstract
This study was designed to evaluate the effects of ethyl pyruvate (EP) on wound healing in primary colonic anastomoses in intraperitoneal sepsis. Standard left colon resection and end-to-end anastomosis were performed on 30 rats. They were grouped as control (C)--no further treatment; sepsis (S)--received 2 mL Escherichia coli (ATCC 25922) intraperitoneally (IP), and after 5 hours, standard resection and anastomosis were performed; or sepsis-group treated with EP (S-EP)--received 2 mL E coli IP, after 5 hours, standard resection and anastomosis were performed and treated with EP 50 mg/kg IP for 7 days. On the postoperative day 7, the animals were sacrificed. The anastomosis bursting pressure in group S was significantly lower than in the other groups. There were no differences between groups C and S-EP. Tissue hydroxyproline concentrations in group C were significantly higher than in group S. EP administration prevented intraperitoneal sepsis-induced impaired anastomotic healing of colon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W245712
Ethyl pyruvate, natural, FG
C5H8O3
E47808
Ethyl pyruvate, 98%
C5H8O3