EMAIL THIS PAGE TO A FRIEND
Geriatrics & gerontology international
3-(2,4-dimethoxybenzylidene)-anabaseine: a promising candidate drug for Alzheimer's disease?
PMID 22300107
Abstract
3-(2,4-dimethoxybenzylidene)-anabaseine is an analog of the paralytic alkaloid, anabaseine, from the ribbon worms Amphiporus sp., that shows numerous properties, in particular an agonist activity on alpha 7 nicotinic acetylcholine receptors. This article reviews these properties and explains to what extent they could be valuable to control symptomatology and/or neurodegeneration in Alzheimer's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0326
GTS-21, ≥97% (HPLC)
C19H20N2O2 · 2HCl