EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Mimicking phosphorylation at serine 87 inhibits the aggregation of human ?-synuclein and protects against its toxicity in a rat model of Parkinson's disease.
PMID 22302797