EMAIL THIS PAGE TO A FRIEND
Ying yong sheng tai xue bao = The journal of applied ecology
[Oat growth and cation absorption characteristics under salt and alkali stress].
PMID 22303664
Abstract
This paper monitored the oat growth and cation absorption characteristics on a saline-alkali soil in the Baicheng region of Jilin Province under low, medium, and high levels of salt stress. No significant differences were observed in the shoot growth and yield components under the three levels of salt stress, but the root biomass and root/shoot ratio decreased significantly with increasing salt stress level. At maturing stage, the root/shoot ratio under medium and high salt stresses was 77.2% and 64.5% of that under low salt stress, respectively. Under the three levels of salt stress, the K+/Na+ and Ca2+/Na+ ratios in oat plant had significant differences at trefoil stage, but no significant differences at heading stage. With the increase of salt stress level, the cation absorption selectivity coefficient of oat at filling stage decreased significantly, but the transportation selectivity coefficient had no significant difference under the three levels of stress. It was concluded that oat could adapt to the salt and alkali stress of soda-alkaline soil to some extent, and the adaptation capability decreased with the increasing level of stress. The decrease of oat root biomass and the stronger ion selective absorption capacity at heading stage under salt and alkali stress could benefit the shoot growth and yield components of oat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
31432
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
71360
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
CNa2O3 · 10H2O
13414
Sodium carbonate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · 10H2O
577782
Sodium carbonate decahydrate, 99.999% trace metals basis
CNa2O3 · 10H2O
S4132
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
CNa2O3 · H2O
13568
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · H2O
230952
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
CNa2O3 · H2O