EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
Estrogen receptor alpha overexpressing mouse antral follicles are sensitive to atresia induced by methoxychlor and its metabolites.
PMID 22306526
Abstract
Methoxychlor (MXC) and its metabolites bind to estrogen receptors (ESRs) and increase ovarian atresia. To test whether ESR alpha (ESR1) overexpressing (ESR1 OE) antral follicles are more sensitive to atresia compared to controls, we cultured antral follicles with vehicle, MXC (1-100 μg/ml) or metabolites (0.1-10 μg/ml). Results indicate that MXC and its metabolites significantly increase atresia in ESR1 OE antral follicles at lower doses compared to controls. Activity of pro-apoptotic factor caspase-3/7 was significantly higher in ESR1 OE treated antral follicles compared to controls. ESR1 OE mice dosed with MXC 64 mg/kg/day had an increased percentage of atretic antral follicles compared to controls. Furthermore, pro-caspase-3 levels were found to be significantly lower in ESR1 OE ovaries than controls dosed with MXC 64 mg/kg/day. These data suggest that ESR1 OE ovaries are more sensitive to atresia induced by MXC and its metabolites in vitro and in vivo compared to controls.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36161
Methoxychlor, PESTANAL®, analytical standard
C16H15Cl3O2
49054
Methoxychlor, analytical standard
C16H15Cl3O2