EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
A femtosecond fluorescence study of vibrational relaxation and cooling dynamics of UV dyes.
PMID 22307066
Abstract
We present a femtosecond broad-band fluorescence up-conversion study of the vibrational relaxation dynamics of two UV chromophores, 2,5-diphenyloxazole (PPO) and para-terphenyl (pTP), pumped with a large excess of vibrational energy (>2000 cm(-1)). The band narrowing of the transient fluorescence spectrum reflects a biphasic cooling process in a few hundreds of fs and a few ps. In the sub-ps regime, our data suggest a structural rearrangement in the excited state, followed by thermalization of the excess energy. These dynamics affect the fluorescence spectra of PPO and pTP in different ways. In PPO, the damping of a low frequency vibrational wavepacket and a significant sub-ps narrowing of the band characterize the vibrational relaxation. In pTP, the latter is faster and appears as a red shift with distortion of the band in <200 fs.