EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
Single dose NTBC-treatment of hereditary tyrosinemia type I.
PMID 22307209
Abstract
NTBC (2-(2-nitro-4-trifluoromethylbenzoyl)-1,3cyclohexanedione) is the mainstay of treatment in tyrosinemia type 1 (HT 1). The current recommendation is to divide the total daily dose of NTBC into two doses. We monitored the plasma NTBC concentrations in a series of seven patients who were changed from multiple divided doses to a single daily dose of NTBC. Two additional patients were started on a single daily dose of NTBC after the diagnosis of HT 1 was established. In three patients, NTBC kinetics were performed over 6 and 24 hours, respectively. The use of multiple divided doses or a single daily dose did not significantly affect plasma NTBC concentrations or the mean daily dose needed to attain therapeutic plasma NTBC concentrations. Moreover, kinetic studies demonstrated that plasma NTBC concentrations were completely stable over a period of 24 hours with a single dose regimen, as expected given the known NTBC plasma half life of 54 hours. Although these preliminary results need to be confirmed in more patients, our findings show that administration of NTBC in a single daily dose may be as effective as a multiple-dose regimen in reaching therapeutic plasma NTBC concentrations and suppressing succinylacetone formation in patients with HT 1. In fact, single dose treatment may increase patients' compliance with the drug treatment and improve metabolic control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1415
4,6-Dioxoheptanoic acid, powder
C7H10O4
SML0269
Nitisinone, ≥95% (HPLC)
C14H10F3NO5