EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Iron-catalysed tandem cross-dehydrogenative coupling (CDC) of terminal allylic C(sp3) to C(sp2) of styrene and benzoannulation in the synthesis of polysubstituted naphthalenes.
PMID 22307234
Abstract
A novel iron-catalysed tandem cross-dehydrogenative coupling and benzoannulation process was developed for the synthesis of biologically and synthetically important polysubstituted naphthalene derivatives from simple 1,2-aryl-propenes and styrenes in moderate to good yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295663
Propylene, ≥99%
C3H6