EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Dipeptidyl peptidase IV (DPPIV/CD26)-based prodrugs of hydroxy-containing drugs.
PMID 22307932
Abstract
We previously described a novel prodrug approach in which a di- or tetrapeptide moiety is linked to a wide variety of amine-containing drugs through an amide bond, which is specifically cleaved by dipeptidyl peptidase IV (DPPIV/CD26) activity. Herein we report the application of this prodrug approach to a variety of hydroxy-containing drugs (primary, secondary, tertiary, or aromatic hydroxy groups). We designed and studied tripartite prodrugs containing a dipeptide moiety (cleavable by DPPIV/CD26) and a valine as a hetero-bifunctional connector to link the dipeptide to the hydroxy group of the drug through a metabolically labile ester bond. The hydroxy-containing prodrugs showed various susceptibilities to hydrolysis by DPPIV/CD26 and serum, depending on the nature of the compound. Prodrugs of compounds containing a primary hydroxy group (as in didanosine) or a hydroxy moiety on an aromatic entity (as in acetaminophen) were most efficiently converted. In contrast, a tertiary hydroxy group was much less susceptible to conversion into its parent drug by DPPIV/CD26 or serum. A number of the prodrugs showed remarkable increases in water solubility relative to their parent drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0162
2′,3′-Dideoxyinosine, ≥98% (HPLC)
C10H12N4O3