EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Microbial biomass and carbon mineralization in agricultural soils as affected by pesticide addition.
PMID 22310842
Abstract
A laboratory study was conducted with four pesticides, viz. a fungicide (carbendazim), two insecticides (chlorpyrifos and cartap hydrochloride) and an herbicide (pretilachlor) applied to a sandy clay loam soil at a field rate to determine their effect on microbial biomass carbon (MBC) and carbon mineralization (C(min)). The MBC content of soil increased with time up to 30 days in cartap hydrochloride as well as chlorpyrifos treated soil. Thereafter, it decreased and reached close to the initial level by 90th day. However, in carbendazim treated soil, the MBC showed a decreasing trend up to 45 days and subsequently increased up to 90 days. In pretilachlor treated soil, MBC increased through the first 15 days, and thereafter decreased to the initial level. Application of carbendazim, chlorpyrifos and cartap hydrochloride decreased C(min) for the first 30 days and then increased afterwards, while pretilachlor treated soil showed an increasing trend.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45995
Cartap hydrochloride, PESTANAL®, analytical standard
C7H15N3O2S2 · HCl
31251
Pretilachlor, PESTANAL®, analytical standard
C17H26ClNO2