EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Identification and characterization of an indigo-producing oxygenase involved in indole 3-acetic acid utilization by Acinetobacter baumannii.
PMID 22311185
Abstract
Acinetobacter baumannii harbours a gene cluster similar to the iac locus of Pseudomonas putida 1290, which can catabolize the plant hormone indole 3-acetic acid (IAA) as an energy source. However, there has been no evidence showing that IAA can be utilized by A. baumannii. This study showed that A. baumannii can grow in M9 minimal medium containing IAA as the sole carbon source. A mutagenesis study indicated that iacA, encoded in the iac locus of A. baumannii, is involved in the catabolism of IAA. As shown by western blotting analysis, the IacA protein was detected in A. baumannii grown in M9 minimal medium with IAA but not with pyruvate, suggesting that the expression of iacA is regulated by the presence of IAA. In vitro studies have shown that IacA can oxidize indole, an IAA-like molecule, converting it to indoxyl, which spontaneously dimerises to form indigo. In this study, we show that the crude extracts from either wild-type A. baumannii or Escherichia coli overexpressing IacA can oxidize IAA. These results imply that the iac gene cluster of A. baumannii is involved in IAA degradation and that the iacA gene is upregulated when cells encounter IAA in their native environments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229296
Indigo, synthetic, Dye content 95 %
C16H10N2O2