EMAIL THIS PAGE TO A FRIEND
The Journal of family practice
Clinical inquiry. Which drugs work best for early Parkinson's disease?
PMID 22312616
Abstract
Levodopa/carbidopa is the most effective medical therapy for Parkinson's disease, but it's associated with dyskinesia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2530
Ropinirole hydrochloride, powder, ≥98% (HPLC)
C16H24N2O · HCl