EMAIL THIS PAGE TO A FRIEND
Terapevticheskii arkhiv
[Perspectives of prevention and treatment of herpes symplex with regular recurrence].
PMID 22312884
Abstract
To assess efficacy of two-stage treatment of severe genital herpes (GH) with regular recurrences: combined use of antivirus medicines with interferon preparations and its inductors followed by antirecurrence vaccine therapy. Two-stage treatment was given to 100 patients suffering from GH with severe regular recurrences. One-stage treatment included famvir in combination with interferon (viferon) or interferon inductors (amixin, cycloferon) and antioxidants. Stage two treatment consisted of vaccine therapy of patients who failed prophylactic standard vaccination by allergometric technique. The first stage treatment prolonged recurrence-free period 2-3-fold in more than 85% patients, improved quality of life. Stage-two treatment resulted in long-term clinicoimmunological remission which is necessary for conduction of anti-recurrence vaccine treatment. Changes in therapeutic-preventive policy in patients with recurrent GH with regular recurrence (2 stage of treatment) prolong recurrence-free intervals, improve quality of life and social adaptation of patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220957
Tilorone dihydrochloride, 95%
C25H34N2O3 · 2HCl