EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
2-(Pyrrolidin-1-yl)ethyl-3,4-dihydroisoquinolin-1(2H)-one derivatives as potent and selective histamine-3 receptor antagonists.
PMID 22313242
Abstract
On the basis of the previously reported benzimidazole 1,3'-bipyrrolidine benzamides (1), a new class of 2-(pyrrolidin-1-yl)ethyl-3,4-dihydroisoquinolin-1(2H)-one derivatives (3-50) were synthesized and evaluated as potent H(3) receptor antagonists. In particular, compound 39 exhibited potent in vitro binding and functional activities at the H(3) receptor, good selectivities against other neurotransmitter receptors and ion channels, acceptable pharmacokinetic properties, and a favorable in vivo profile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I28208
Isoquinoline, 97%
C9H7N
282162
Isoquinoline, technical grade, 90-92%
C9H7N