EMAIL THIS PAGE TO A FRIEND
The Kaohsiung journal of medical sciences
Effect of the endothelin receptor antagonist tezosentan on alpha-naphthylthiourea-induced lung injury in rats.
PMID 22313533
Abstract
Acute lung injury is an inflammatory syndrome that increases the permeability of the blood-gas barrier, resulting in high morbidity and mortality. Despite intensive research, treatment options remain limited. We investigated the protective efficacy of tezosentan, a novel, dual endothelin receptor antagonist, in an experimental model of alpha-naphthylthiourea (ANTU)-induced acute lung injury in rats. ANTU was intraperitoneally (i.p.) injected into rats at a dose of 10 mg/kg. Tezosentan was injected 30 minutes before ANTU was subcutaneously (s.c.) injected at doses of 2, 10, or 30 mg/kg, 60 minutes before ANTU was injected at doses of 2, 10, or 30 mg/kg (i.p.), and 90 minutes before ANTU at a dose of 10 mg/kg (i.p.). Four hours later, the lung weight/body weight (LW/BW) ratio and pleural effusion (PE) were measured. When injected 30 minutes before ANTU at doses of 2, 10, or 30 mg/kg (s.c.), tezosentan had no effect on lung pathology. When injected 60 minutes before ANTU at doses of 2, 10, or 30 mg/kg (i.p.) or 90 minutes before ANTU (10 mg/kg, i.p.), tezosentan significantly decreased the PE/BW ratio and had a prophylactic effect on PE formation at all doses. Therefore, tezosentan may attenuate lung injury. Furthermore, its acute and inhibitory effects on fluid accumulation were more effective in the pleural cavity than in the interstitial compartment in this experimental model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45328
ANTU, PESTANAL®, analytical standard
C11H10N2S