EMAIL THIS PAGE TO A FRIEND
The Prostate
miR-141 modulates androgen receptor transcriptional activity in human prostate cancer cells through targeting the small heterodimer partner protein.
PMID 22314666
Abstract
Aberrant expressions of microRNAs, including upregulation of miR-141, are closely associated with the tumorigenesis of prostate cancer (PCa). The orphan receptor small heterodimer partner (Shp) is a co-repressor to androgen receptor (AR) and represses AR-regulated transcriptional activity. Here, we investigated the correlation of Shp expression with the cellular level of miR-141 and its effects on AR transcriptional activity in non-malignant and malignant human prostate epithelial cell lines. We found that Shp was downregulated in multiple PCa cell lines. The mature form of miR-141 was upregulated in PCa cells. miR-141 could target 3'-untranslated region of Shp mRNA resulting in translational suppression and RNA degradation. Moreover, enforced expression of Shp or inhibition of miR-141 function by anti-miR-141 attenuated AR-regulated transcriptional activity in AR-responsive LNCaP cells. Phenethyl isothiocyanate, a natural constituent of many edible cruciferous vegetables, increased Shp expression, downregulated miR-141, and inhibited AR transcriptional activity in LNCaP cells. Shp is a target for miR-141 and it is downregulated in cultured human PCa cells with the involvement of upregulation of miR-141, which promotes AR transcriptional activity. Moreover, Shp and miR-141 could be targets for chemoprevention for PCa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W401404
2-Phenylethyl isothiocyanate, FG
C9H9NS
253731
Phenethyl isothiocyanate, 99%
C9H9NS