EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Contaminated drugs are held responsible for 120 deaths in Pakistan.
PMID 22315298
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0775010 Isosorbide mononitrate, European Pharmacopoeia (EP) Reference Standard
C6H9NO6