EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health
Neurotoxicity and carcinogenicity of N-methylolacrylamide in F344 rats and B6C3F1 mice.
PMID 2231776
Abstract
Toxicology and carcinogenicity studies of N-methylolacrylamide were conducted by administering the chemical by gavage in water to both sexes of F344/N rats and B6C3F1 mice 5 times per week for 16 d, 13 wk, or 2 yr. In 16-d studies, rats receiving doses of 200 mg/kg or higher and mice receiving 400 mg/kg died. In 13-wk studies, all rats given 100 mg/kg or higher doses died. Rats receiving 50 mg/kg or higher doses developed hindlimb ataxia progressing to paralysis. In neurobehavioral assessments, decreased forelimb and hindlimb grip strength occurred in rats at doses as low as 12.5 mg/kg. Landing footspread was also increased in dosed rats compared to controls. Axon filament and myelin sheath degeneration in the spinal cord and/or peripheral nerves occurred in rats receiving doses of 25 mg/kg or higher. Necrosis in the granular cell layer of the cerebellum was seen in rats given 200 mg/kg. Mice receiving 200 mg/kg in 13-wk studies died. Decreased grip strength was noted in mice at doses as low as 25 mg/kg, and rotarod performance was also affected by N-methylolacrylamide administration, but no neuropathology was seen microscopically. Testicular weights were decreased at doses as low as 12.5 mg/kg, and hepatocellular necrosis, thymic lymphocyte necrosis, and hemorrhage, necrosis, and mineralization of the zona reticularis of the adrenal gland were seen in mice that died (200 mg/kg). In 2-yr studies, survival and weight gains in male and female rats receiving doses of 6 or 12 mg/kg/d were minimally affected. No biologically important clinical signs or neoplastic or nonneoplastic lesions were attributed to N-methylolacrylamide administration to rats, suggesting that higher doses could have been tolerated. In mice, survival was not different between dosed and control groups (0, 25, or 50 mg/kg/d). Body weights were higher by as much as 25% in dosed compared to control groups. No compound-related clinical signs were observed, but increases in neoplasms of the harderian gland, liver, and lung were clearly related to chemical administration in both sexes of mice. Benign granulosa-cell neoplasms of the ovary were also increased in dosed female mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245801
N-(Hydroxymethyl)acrylamide solution, 48 wt. % in H2O
C4H7NO2