EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Tubulin binding, protein-bound conformation in solution, and antimitotic cellular profiling of noscapine and its derivatives.
PMID 22320354
Abstract
Noscapine and its 7-hydroxy and 7-amino derivatives were characterized for their binding to tubulin. A solution NMR structure of these compounds bound to tubulin shows that noscapine and its 7-aniline derivative do not compete for the same binding site nor does its small molecule crystal structure match its tubulin-bound conformation. These compounds were also tested for their antiproliferative effects on a panel hepatocellular carcinoma cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
363960
(S,R)-Noscapine, 97%
C22H23NO7