EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Determination of histamine in seafood by hydrophilic interaction chromatography/tandem mass spectrometry.
PMID 22322812
Abstract
A simple method was developed to determine histamine, an important compound in chemical food poisoning, by extraction followed by hydrophilic interaction chromatography-tandem mass spectrometry using a hydrophilic column with sulfobetaine-type zwitterion groups. The quantitation range in seafood products was from 0.4 to 200 mg kg(-1) for 5 g food samples. Quantitative recoveries were obtained with four types of seafood product. These results agreed well with those from the more complex, conventional HPLC method, which requires sample derivatization with dansyl chloride.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S