EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Appetite suppressant was probably responsible for 1300 deaths, study shows.
PMID 22323511
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7522
Benfluorex hydrochloride, analytical standard
C19H20F3NO2 · HCl