EMAIL THIS PAGE TO A FRIEND
Journal of Korean medical science
Tandem high-dose chemotherapy and autologous stem cell transplantation in young children with atypical teratoid/rhabdoid tumor of the central nervous system.
PMID 22323859
Abstract
The feasibility and effectiveness of tandem high-dose chemotherapy and autologous stem cell transplantation (HDCT/autoSCT) were evaluated in children younger than 3 yr of age with atypical teratoid/rhabdoid tumors (ATRT). Tandem HDCT/autoSCT was administered following six cycles of induction chemotherapy. Radiotherapy (RT) was administered if the tumor relapsed or progressed, otherwise, it was administered after 3 yr of age. Tumors relapsed or progressed during induction chemotherapy in 5 of 9 patients enrolled; 3 of these 5 received tandem HDCT/autoSCT as a salvage treatment. One patient died from sepsis during induction chemotherapy. The remaining 3 patients proceeded to tandem HDCT/autoSCT; however, 2 of these patients showed tumor relapse/progression after tandem HDCT/autoSCT. All 7 relapses/progressions occurred at primary sites even in patients with leptomeningeal seeding. Toxicities during tandem HDCT/autoSCT were manageable. A total of 5 patients were alive with a median follow-up of 20 (range 16-70) months from diagnosis. Four of 5 patients who received RT after relapse/progression are alive. The probability of overall survival at 3 yr from diagnosis was 53.3% ± 17.3%. Our tandem HDCT/autoSCT is feasible; however, early administration of RT prior to tandem HDCT/autoSCT should be considered to improve the outcome after tandem HDCT/autoSCT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6069
Thio-TEPA, 98%, solid
C6H12N3PS