EMAIL THIS PAGE TO A FRIEND
ACS chemical biology
Amide analogues of CD1d agonists modulate iNKT-cell-mediated cytokine production.
PMID 22324848
Abstract
Invariant natural killer T (iNKT) cells are restricted by the non-polymorphic MHC class I-like protein, CD1d, and activated following presentation of lipid antigens bound to CD1d molecules. The prototypical iNKT cell agonist is α-galactosyl ceramide (α-GalCer). CD1d-mediated activation of iNKT cells by this molecule results in the rapid secretion of a range of pro-inflammatory (Th1) and regulatory (Th2) cytokines. Polarization of the cytokine response can be achieved by modifying the structure of the glycolipid, which opens up the possibility of using CD1d agonists as therapeutic agents for a range of diseases. Analysis of crystal structures of the T-cell receptor-α-GalCer-CD1d complex led us to postulate that amide isosteres of known CD1d agonists should modulate the cytokine response profile upon iNKT-cell activation. To this end, we describe the synthesis and biological activity of amide analogues of α-GalCer and its non-glycosidic analogue threitol ceramide (ThrCer). All of the analogues were found to stimulate murine and human iNKT cells by CD1d-mediated presentation to varying degrees; however, the thioamide and carbamate analogues of ThrCer were of particular interest in that they elicited a strongly polarized cytokine response (more interferon-gamma (IFN-γ), no interleukin-4 (IL-4)) in mice. While the ThrCer-carbamate analogue was shown to transactivate natural killer (NK) cells, a mechanism that has been used to account for the preferential production of IFN-γ by other CD1d agonists, this pathway does not account for the polarized cytokine response observed for the thioamide analogue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
377619
D-Threitol, 99%
C4H10O4
263559
DL-Threitol, 97%
C4H10O4