EMAIL THIS PAGE TO A FRIEND
International journal of rheumatic diseases
Advanced glycation end-products inhibition improves endothelial dysfunction in rheumatoid arthritis.
PMID 22324946
Abstract
Chronic inflammation in rheumatoid arthritis is associated with vascular endothelial dysfunction. The objective was to study the efficacy and safety of advanced glycation end products (AGEs) inhibitor (benfotiamine 50 mg + pyridoxamine 50 mg + methylcobalamin 500 μg, Vonder(®) (ACME Lifescience, Baddi, Himachal Pradesh, India)) on endothelial function in rheumatoid arthritis (RA). Twenty-four patients with established active RA with high disease activity (Disease Activity Score of 28 joints [DAS28 score] > 5.1) despite treatment with stable doses of conventional disease-modifying antirheumatic drugs were investigated. Inflammatory disease activity (DAS28 and Health Assessment Questionnaire-Disability Index [HAQ-DI] scores, erythrocyte sedimentation rate [ESR] and C-reactive protein [CRP]), markers of endothelial dysfunction, serum nitrite concentration and endothelium-dependent and -independent vasodilation of the brachial artery were measured before and after 12 weeks therapy with twice a day oral AGEs inhibitor. After treatment, flow-mediated vasodilation improved from 9.64 ± 0.65% to 15.82 ± 1.02% (P < 0.01), whereas there was no significant change in endothelium-independent vasodilation with nitroglycerin and baseline diameter; serum nitrite concentration significantly reduced from 5.6 ± 0.13 to 5.1 ± 0.14 μmol/L (P = 0.004), ESR from 63.00 ± 3.5 to 28.08 ± 1.5 mm in the first h (P < 0.01) and CRP levels from 16.7 ± 4.1 to 10.74 ± 2.9 mg/dL (P < 0.01). DAS28 and HAQ-DI scores were significantly reduced, from 5.9 ± 0.17 to 3.9 ± 0.17 (P < 0.01) and 4.6 ± 0.17 to 1.7 ± 0.22 (P < 0.01), respectively. Advanced glycation end products inhibitor improves endothelial dysfunction and inflammatory disease activity in RA. In RA, endothelial dysfunction is part of the disease process and is mediated by AGEs-induced inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9756
Methylcobalamin, vitamin B12 analog
C63H91CoN13O14P
B9636
S-Benzoylthiamine O-monophosphate
C19H23N4O6PS