EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Degradation of carbofuran derivatives in restricted water environments: basic hydrolysis in AOT-based microemulsions.
PMID 22325067
Abstract
The effect of sodium bis(2-ethylhexyl)sulfosuccinate/isooctane/water microemulsions on the stability of 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methylcarbamate (carbofuran, CF), 3-hydroxy-2,3-dihydro-2,2-dimethylbenzofuran-7-yl methylcarbamate (3-hydroxycarbofuran, HCF) and 3-keto-2,3-dihydro-2,2-dimethylbenzofuran-7-yl methylcarbamate (3-ketocarbofuran, KCF) in basic media has been studied. The presence of these microheterogeneous media implies a large basic hydrolysis of CF and HCF on increasing surfactant concentration and, also, on increasing water content in the microemulsion. The hydrolysis rate constants are approximately 2- and 10-fold higher than those in pure water for HCF and CF, respectively. In contrast, a steep descent in the rate of decomposition for KCF was observed. These behaviours can be ascribed to the presence of CF derivatives both in the hydrophilic phase and in the lipophilic phase, while the hydroxyl ions are only restricted to the water pool of the microemulsion (hydrophilic phase). The kinetic rate constants for the basic hydrolysis in AOT-based microemulsions have been obtained on the basis of a pseudophase model. Taking into account that an important part of soils are colloids, the possibility of the presence of restricted water environments implies that soil composition and its structure will play an important role in the stability of these carbamates. In fact, we observed that the presence of these restricted aqueous media in the environment, in particular in watersheds and in wastewaters, could reduce significantly the half-life of these pesticides (33% and 91% for HCF and CF, respectively).