EMAIL THIS PAGE TO A FRIEND
Veterinary immunology and immunopathology
Potential of auxotrophic Edwardsiella tarda double-knockout mutant as a delivery vector for DNA vaccine in olive flounder (Paralichthys olivaceus).
PMID 22325745
Abstract
To evaluate potential of an auxotrophic Edwardsiella tarda mutant (Δalr Δasd E. tarda) as a delivery vehicle for DNA vaccine in fish, olive flounder (Paralichthys olivaceus) were immunized with the E. tarda mutant harboring plasmids (pG02-ASD-CMV-eGFP) for eukaryotic expression of the enhanced green fluorescent protein (eGFP) gene through either intraperitoneal (i.p.) or oral route, and the expression of eGFP in the internal organs and generation of antibody against eGFP in fish were analyzed. In fish i.p. injected with 2×10(7)CFU/fish of Δalr Δasd E. tarda harboring pG02-ASD-CMV-eGFP, expression of eGFP was detected in liver, kidney, and spleen from 1 day to 28 days post-injection. In fish orally administered with 1×10(9)CFU/fish of the bacteria, the eGFP band was detected in liver, kidney, and spleen from 1 day to 14 days post-administration, whereas, in intestine, the band was detected only at 1 day post-administration. Either oral or i.p. immunization of olive flounder with recombinant E. tarda that carried eGFP-expressing eukaryotic plasmids was successful to induce humoral adaptive immunity against not only E. tarda that was used as a delivery vehicle but also eGFP that was used as the reporter protein of DNA vaccine, suggesting attenuated E. tarda-vectored DNA vaccine has a potential to be used as a combined vaccine against infectious diseases in fish.