EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
A combination of 2-deoxy-D-glucose and 6-aminonicotinamide induces cell cycle arrest and apoptosis selectively in irradiated human malignant cells.
PMID 22328137
Abstract
Previously, we have shown that a combination of metabolic modifiers 2-deoxy-D-glucose (2-DG) and 6-aminonicotinamide (6-AN) results in oxidative stress mediated radiosensitization of malignant cells via mitochondrial dysfunction and non-coordinated expression of antioxidant defense, besides inhibition of repair and recovery. In the present study, our objective was to study, in a panel of human malignant cells of various origins (lung carcinoma, squamous carcinoma, oral carcinoma, and glioblastoma), if the inhibitory activity of combination (2-DG+6-AN+2 Gy) against tumor growth could be considered a general phenomenon and to determine its effect on the cell cycle. The results revealed that combination (2-DG+6-AN+2 Gy) treatment result in significant cell growth inhibition and induced ROS generation in all cancer cells studied. The anti-proliferative effect was related to the ability of combination (2-DG+6-AN+2 Gy) to provoke growth inhibition at the G2/M arrest and apoptosis. Furthermore, combination (2-DG+6-AN+2 Gy) induced G2/M arrest is closely correlated to decreased cyclin A, cyclin B1, and cdc2 levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A68203
6-Aminonicotinamide, 99%
C6H7N3O