EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Research
The unliganded glucocorticoid receptor positively regulates the tumor suppressor gene BRCA1 through GABP beta.
PMID 22328717
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, clone, from rabbit