EMAIL THIS PAGE TO A FRIEND
TSitologiia i genetika
[Polymorphism of endocellular isoenzymes Schizophyllum commune Fr. (Basidiomycetes) in territory of Donetsk area].
PMID 22329162
Abstract
Results of research of endocellular isoenzyme polymorphism of Schizophyllum commune Fr. cultures growing on the territory of Donetsk region are presented. Description of AMY, ADH, GPDH, GDH, SDH, EST electrophoretic spectrum has been carried out. The enzyme systems ADH, GPDH and GDH were monomorphic. The greatest variety of endocellular isoforms was shown for EST. Well defined six zones were detected and for three of them the polymorphism was peculiar.