EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Site specific chemoselective labelling of proteins with robust and highly sensitive Ru(II) bathophenanthroline complexes.
PMID 22331141
Abstract
The bioorthogonal and chemoselective fluorescence labelling of several cell-free synthesized proteins containing a site-specifically incorporated azido amino acid was possible using different alkyne-functionalized Ru(II) bathophenanthroline complexes. We were able to achieve a selective labelling even in complex mixtures of proteins despite the fact that ruthenium dyes normally show a high tendency for unspecific interactions with proteins and are commonly used for total staining of proteins. Since the employed Ru complexes are extremely robust, photo-stable and highly sensitive, the approach should be applicable to the production of labelled proteins for single molecule spectroscopy and fluorescence-based interaction studies.
Related Materials