EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Medicating feeds for early lambing ewes.
PMID 22331786
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33823
Decoquinate, VETRANAL, analytical standard
C24H35NO5