EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Mechanistic studies on the Pd-catalyzed direct C-H arylation of 2-substituted thiophene derivatives with arylpalladium bipyridyl complexes.
PMID 22334438
Abstract
Direct C-H phenylation of 2-ethylthiophene and 2-chlorothiophene with PhPdI(bipy) complex to form either the corresponding 4-phenyl or 5-phenylthiophene derivative is studied under stoichiometric conditions using various Lewis acids as additives. It is shown that reactions occur via the corresponding cationic Pd complex (PhPdbipy(+)) and that the counteranion determines the regioselectivity. High-level DFT calculations reveal that C-C bond formation occurs via a carbopalladation pathway and not via electrophilic palladation. These calculations give some indications regarding the regioselectivity of the thiophene arylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
155667
2-Chlorothiophene, 96%
C4H3ClS