EMAIL THIS PAGE TO A FRIEND
Vestnik otorinolaringologii
[The results of adjuvant therapy of juvenile recurring respiratory papillomatosis with the use of indole-3-carbinol].
PMID 22334925
Abstract
The authors analyse the efficacy of anti-relapse therapy of juvenile recurring respiratory papillomatosis in 87 children aged from 2 to 15 years with the use of indole-3-carbinol. Prior to inclusion into this study, the patients underwent from 2 to 86 (mean 12 +/- 14) surgical interventions for the ablation of papillomas. The average interval between successive relapses of papillomas ranged between 2 weeks and 12 months (mean 4.9 +/- 2.33 months). The patients remained under observation in the clinic during 2 years--6 years 5 months (mean 44.8 +/- 15.93 months). The duration of therapy with indole-3-carbinol varied from 12 weeks to 2 years (mean 8.9 +/- 4.72 months). The stable remission of pathology was documented in 28.7% of the patients within the 2 to 6 year follow-up period. A significant (1.5-10-fold) increase in the duration of interval between successive relapses occurred in 41.1% of the children. In 29.9% of the patients, therapy produced no apparent clinical effect; 18.4% of them showed an insignificant shortening of the interval between relapses that remained unaltered in the remaining 11.5%. No adverse effects of the treatment were recorded. It is concluded that treatment with indole-3-carbinol can be recommended as a starting therapeutic modality for the management of juvenile recurring respiratory papillomatosis and the reduction of the intervals between relapses of the disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7256
Indole-3-carbinol
C9H9NO