EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
MC1R variant allele effects on UVR-induced phosphorylation of p38, p53, and DDB2 repair protein responses in melanocytic cells in culture.
PMID 22336944
Abstract
Variant alleles of the human melanocortin 1 receptor (MC1R) reduce the ability of melanocytes to produce the dark pigment eumelanin, with R alleles being most deficient. Cultured melanocytes of MC1R R/R variant genotype give reduced responses to [Nle(4), D-Phe(7)]α-melanocyte-stimulating hormone (NDP-MSH) ligand stimulation and lower levels of DNA repair than MC1R wild-type strains. p38 controls xeroderma pigmentosum (XP)-C recruitment to DNA damage sites through regulating ubiquitylation of the DNA damage-binding protein 2 (DDB2) protein, and p53 is implicated in the nuclear excision repair process through its regulation of XP-C and DDB2 protein expression. We report the effects of MC1R ligand treatment and UVR exposure on phosphorylation of p38 and p53, and DDB2 protein expression in MC1R variant strains. Wild-type MC1R melanocyte strains grown together with keratinocytes in coculture, when treated with NDP-MSH and exposed to UVR, gave synergistic activation of p38 and p53 phosphorylation, and were not replicated by R/R variant melanocytes, which have lower basal levels of phosphorylated forms of p38. Minor increases in p38 phosphorylation status in R/R variant melanocyte cocultures could be attributed to the keratinocytes alone. We also found that MC1R wild-type strains regulate DDB2 protein levels through p38, but MC1R R/R variant melanocytes do not. This work confirms the important functional role that the MC1R receptor plays in UVR stress-induced DNA repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8764
[Nle4, D-Phe7]-α-Melanocyte Stimulating Hormone trifluoroacetate salt, ≥95% (HPLC)
C78H111N21O19 · xC2HF3O2