EMAIL THIS PAGE TO A FRIEND
Journal of analytical toxicology
Analytical evaluation of four on-site oral fluid drug testing devices.
PMID 22337784
Abstract
The use of oral fluid (OF) as an alternative matrix for the detection of drugs of abuse has increased over the last decade, leading to the need for a rapid, simple, and reliable on-site OF testing device. Four on-site OF drug testing devices (Dräger DrugTest 5000, Cozart DDS, Mavand Rapid STAT, and Innovacon OrAlert) were evaluated on 408 volunteers at drug treatment centers. UPLC-MS-MS results were used as reference to determine sensitivity, specificity and accuracy for each device, applying Belgian legal confirmation cutoffs for benzoylecgonine, cocaine, and THC (10 ng/mL); morphine and 6-acetylmorphine (5 ng/mL); and amphetamine and 3,4-methylenedioxymethylamphetamine (25 ng/mL). Sensitivity for cocaine was 50%, 50%, 27%, and 11% for DrugTest, OrAlert, Rapid STAT, and DDS 806, respectively. For opiates, sensitivities were 84%, 73%, 77%, and 65%, respectively. For THC, the sensitivities were 81%, 23%, 43%, and 28%, respectively. For amphetamines, the sensitivities were 75%, 33%, 17%, and 67%, respectively. Specificity was >88% for opiates and THC, > 90% for amphetamines, and > 97% for cocaine. All tests showed good specificity. DrugTest had the highest sensitivity, although it was still low for some analytes.