EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Estimation of maximal cortisol secretion rate in healthy humans.
PMID 22337905
Abstract
Cortisol secretion is related to ACTH concentration by a sigmoidal dose-response curve, in which high ACTH concentrations drive maximal cortisol secretion rates (CSR(max)). We sought to estimate CSR(max) and free cortisol half-life in healthy humans (n = 21) using numerical methods applied to data acquired during cosyntropin (250 μg) stimulation. We also evaluated the effect of overnight dexamethasone (DEX; 1 mg) vs. placebo on estimates of CSR(max) and free cortisol half-life. This study was a double-blind, placebo-controlled, randomized order of overnight DEX vs. placebo, cosyntropin (250 μg) stimulation with frequent serum cortisol sampling and computer-assisted numerical analysis. The study was conducted at a single academic medical center. Twenty-one healthy adult subjects (15 females and six males), mean aged 46 yr, participated in the study. Intervention in the study included DEX vs. placebo pretreatment, cosyntropin (250 μg) iv with frequent cortisol sampling. CSR(max) and free cortisol half-life estimates, R² for goodness of fit, were measured. Mean ± sd CSR(max) was 0.44 ± 0.13 nm/second, with free cortisol half-life of 2.2 ± 1.1 min. DEX did not significantly affect estimates of CSR(max) or free cortisol half-life. Our model accounts for most of the variability of measured cortisol concentrations (overall R² = 90.9 ±11.0%) and was more accurate (P = 0.004) during DEX suppression (R² = 94.6 ± 4.6%) compared with placebo (R² = 87.2 ± 8.7%). Application of a mass-action model under conditions of cosyntropin stimulation provides a relatively simple method for estimation CSR(max) that accurately predicts measured cortisol concentrations. DEX administration did not significantly affect estimates of CSR(max) or free cortisol half-life.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0298
Adrenocorticotropic Hormone Fragment 1-24 human, rat, ≥97% (HPLC)
C136H210N40O31S