EMAIL THIS PAGE TO A FRIEND
European review for medical and pharmacological sciences
Anti-Candida activity and brine shrimp toxicity assay of Ganoderma boninense.
PMID 22338547
Abstract
Ganoderma (G.) boninense is a white rot fungus, which can be found in the palm oil tree. Several studies have shown that G. boninense has antimicrobial and antagonistic properties. However, there is limited information reported on antifungal properties especially on Candida (C) albicans. Hence, this study was conducted to determine the anti-Candida activity of G. boninense against C albicans. Crude methanolic extracts of G. boninense was obtained by maceration method with 70% methanol. Anti-Candida test was carried out using disc diffusion assay, broth dilution method, time killing profile and brine shrimp toxicity assay. Anti-Candida activity indicated that the mean zone of inhibition was 12.5 +/- 0.6 mm. The MIC value for C. albicans found to be 3.125 mg/ml. The result from time-killing profile showed that the growth of C albicans was inhibited hence decreases its exponential phase. For brine shrimp toxicity assay, the LC50 value was 3.59 mg/ml which proved that the extract of G. boninense is not toxic.
Related Materials