EMAIL THIS PAGE TO A FRIEND
Organic letters
Catalytic enantioselective desymmetrization of meso-glutaric anhydrides using a stable Ni2-Schiff base catalyst.
PMID 22339049
Abstract
We describe the desymmetrization of meso-glutaric anhydrides to chiral hemiesters using a bench-stable homodinuclear Ni(2)-(Schiff base) complex as the catalyst in good to excellent yield (up to 99%) and enantioselectivity (up to 94%). Using the opposite enantiomer of the catalyst, we obtained the same yield and enantioselectivity with the opposite configuration, thereby gaining access to both hemiester enantiomers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3806
Glutaric anhydride, 95%
C5H6O3