EMAIL THIS PAGE TO A FRIEND
International journal of pediatric otorhinolaryngology
Sodium tetradecyl sulphate sclerotherapy for treating venous malformations of the oral and pharyngeal regions in children.
PMID 22341629
Abstract
Venous vascular malformations in the head and neck region present a difficult management challenge. We describe our experience of using sodium tetradecyl sulphate injection sclerotherapy to treat children presenting with venous malformations of the oral and pharyngeal region. We performed a retrospective case note review of consecutive children treated at our institution between 2004 and 2011. Patient notes were analysed for demographic details, site and size of lesion, number and duration of treatments, treatment response and complications. Twelve patients were included (7 boys and 5 girls, mean age 7 years). Sites of lesions included tongue, floor of mouth, pharynx, tonsillar fossae, parapharyngeal space and soft palate. All patients were treated with 3% sodium tetradecyl sulphate (STS) foam injected trans-orally or percutaneously under ultrasound or fluoroscopic guidance. The lesions had a mean volume of 4 ml (range 2-14 ml). An average of 3 treatments was required (range 1-9). In 4 patients a single treatment was sufficient. For those patients requiring multiple treatments, a mean of 4 treatments were required over an average period of 28 months. The overall response rate was 83% (10/12). Complete resolution was achieved in 4 cases (33%) with a significant reduction in size in a further 6 cases (50%). Larger lesions generally require more treatments than low volume lesions. Two cases recurred despite treatment. One patient suffered minor bleeding following transcutaneous injection. Injection sclerotherapy using STS foam offers an effective treatment option when managing children presenting with venous malformations in the oral and pharyngeal. A single treatment may be adequate for small lesions but the procedure may be safely repeated until a satisfactory result is obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293938
Tetradecyl sulfate sodium salt, 95%
C14H29NaO4S