EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
trans-1,2-Diaminocyclohexane mesoporous silica for asymmetric catalysis: enhancement of chirality through confinement space by the plug effect.
PMID 22344411
Abstract
trans-1,2-Diaminocyclohexane functionalized mesoporous silica was applied as an ideal catalyst for asymmetric Michael addition of various nitroalkane derivatives. Short channels and plugs in the pore structure offered chiral enhancement in Michael addition.