EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Stereoselective degradation of metalaxyl and its enantiomers in rat and rabbit hepatic microsomes in vitro.
PMID 22348420
Abstract
The stereoselective degradations of racemate metalaxyl (rac-MX) and its single enantiomers in rat and rabbit hepatic microsomes were assayed by a chiral high-performance liquid chromatography method. The t(1/2) of (+)-S-MX in rat liver microsomes was between 7-8 min tested by rac-MX and the individual (+)-S-enantiomer, respectively, and that for (-)-R-MX was 15-16 min. In contrast, t(1/2) in rabbit liver microsomes was much longer and showed great difference when using racemate and single enantiomer, which was similar to the results of in vivo study. The enantioselectivity in rat hepatic microsomes was more evident and the degradations of MX enantiomers in rat and rabbit hepatic microsomes were Nicotinamide adenine dinucleotide phosphate-dependent. Michaelis constant (K(m)) and intrinsic metabolic clearance (CL(int)) of (+)-S-MX were larger than that of (-)-R-MX and there was no chiral inversion from (+)-S-MX to (-)-R-MX or vice versa in both rat and rabbit hepatic microsomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N12380
Metalaxyl, analytical standard
C15H21NO4
32012
Metalaxyl, PESTANAL®, analytical standard
C15H21NO4